CH4102149780 ChemBridge Corporation

1-(5-chloropyridin-2-yl)piperazine

  • MDL number MFCD04115049
  • Cas Number : 87394-65-8
  • Molecular Weight : 197.6647

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004469

Safety Information

MSDS https://www.hit2lead.com/sds/4004469_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart