CH4102149869 ChemBridge Corporation

3-acetyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD05864511
  • Cas Number : 1297537-45-1
  • Molecular Weight : 154.1234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4005123

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4005123
Availability details for
Quantity
  • Availability
  • Add to Cart