CH4102149904 ChemBridge Corporation

5-methoxy-2-furoic acid

  • MDL number MFCD02736493
  • Cas Number : 94084-62-5
  • Molecular Weight : 142.1094

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4001967

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4001967
Availability details for
Quantity
  • Availability
  • Add to Cart