CH4102149908 ChemBridge Corporation

1-(7-amino-2,3-dihydro-1,4-benzodioxin-6-yl)ethanone

  • MDL number MFCD00561825
  • Cas Number : 164526-13-0
  • Molecular Weight : 193.1992

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4005993

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4005993
Availability details for
Quantity
  • Availability
  • Add to Cart