CH4102149959 ChemBridge Corporation

4-(2-fluorophenoxy)aniline

  • MDL number MFCD02663348
  • Cas Number : 305801-12-1
  • Molecular Weight : 203.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4009239

Safety Information

MSDS https://www.hit2lead.com/sds/4009239_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart