CH4102149996 ChemBridge Corporation

1-(1-isopropylpiperidin-3-yl)-N-methylmethanamine

  • MDL number MFCD07643226
  • Cas Number : 876716-01-7
  • Molecular Weight : 170.2951

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4009638

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4009638
Availability details for
Quantity
  • Availability
  • Add to Cart