CH4102150003 ChemBridge Corporation

5-methoxy-3-methyl-1H-indole-2-carbaldehyde

  • MDL number MFCD06011082
  • Cas Number : 30464-90-5
  • Molecular Weight : 189.2105

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4009670

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4009670
Availability details for
Quantity
  • Availability
  • Add to Cart