CH4102150004 ChemBridge Corporation

5-fluoro-3-methyl-1H-indole-2-carbaldehyde

  • MDL number MFCD06011083
  • Cas Number : 842972-09-2
  • Molecular Weight : 177.175

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4009671

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4009671
Availability details for
Quantity
  • Availability
  • Add to Cart