CH4102150015 ChemBridge Corporation

(3-cyano-2-methyl-1H-indol-1-yl)acetic acid

  • MDL number MFCD03216644
  • Cas Number : 531544-17-9
  • Molecular Weight : 214.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4009759

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/building-blocks/4009759
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart