CH4102150036 ChemBridge Corporation

4-amino-N-methyl-1,2,5-oxadiazole-3-carboxamide

  • MDL number MFCD08059796
  • Cas Number : 30720-84-4
  • Molecular Weight : 142.116

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4009847

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4009847
Availability details for
Quantity
  • Availability
  • Add to Cart