CH4102150076 ChemBridge Corporation

(4-methyl-1,2,5-oxadiazol-3-yl)acetic acid

  • MDL number MFCD06010348
  • Cas Number : 15323-69-0
  • Molecular Weight : 142.1127

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010224

Safety Information

MSDS https://www.hit2lead.com/sds/4010224_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart