CH4102150099 ChemBridge Corporation

3-(3-pyridin-3-yl-1,2,4-oxadiazol-5-yl)propanoic acid

  • MDL number MFCD07186410
  • Cas Number : 876716-11-9
  • Molecular Weight : 219.1968

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010287

Safety Information

MSDS https://www.hit2lead.com/sds/4010287_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart