CH4102150124 ChemBridge Corporation

2-hydroxy-5-(1H-tetrazol-1-yl)benzoic acid

  • MDL number MFCD00826248
  • Cas Number : 339310-80-4
  • Molecular Weight : 206.1582

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010369

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4010369
Availability details for
Quantity
  • Availability
  • Add to Cart