CH4102150125 ChemBridge Corporation

4-(2-methyl-1H-imidazol-1-yl)aniline

  • MDL number MFCD06797788
  • Cas Number : 74852-81-6
  • Molecular Weight : 173.2144

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010370

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4010370_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart