CH4102150148 ChemBridge Corporation

2-(2-oxopyrrolidin-1-yl)propanoic acid

  • MDL number MFCD05237228
  • Cas Number : 67118-32-5
  • Molecular Weight : 157.1671

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010424

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4010424
Availability details for
Quantity
  • Availability
  • Add to Cart