CH4102150197 ChemBridge Corporation

N-ethyl-3-morpholin-4-ylpropan-1-amine

  • MDL number MFCD08691500
  • Cas Number : 915921-49-2
  • Molecular Weight : 172.2679

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010712

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4010712
Availability details for
Quantity
  • Availability
  • Add to Cart