CH4102150199 ChemBridge Corporation

2-(4-fluorophenyl)piperidine

  • MDL number MFCD02663556
  • Cas Number : 383128-03-8
  • Molecular Weight : 179.234

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4002376

Safety Information

MSDS https://www.hit2lead.com/sds/4002376_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart