CH4102150210 ChemBridge Corporation

1-cyclopropyl-5-oxopyrrolidine-3-carboxylic acid

  • MDL number MFCD06660782
  • Cas Number : 876716-43-7
  • Molecular Weight : 169.1778

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010746

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4010746
Availability details for
Quantity
  • Availability
  • Add to Cart