CH4102150617 ChemBridge Corporation

3-(2,5-dimethylphenyl)-1H-pyrazole-4-carbaldehyde

  • MDL number MFCD08059872
  • Cas Number : 720696-71-9
  • Molecular Weight : 200.2365

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012160

Safety Information

MSDS https://www.hit2lead.com/sds/4012160_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart