CH4102150733 ChemBridge Corporation

3-benzyl-1H-1,2,4-triazol-5-amine

  • MDL number MFCD00465604
  • Cas Number : 22819-07-4
  • Molecular Weight : 174.2025

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4015723

Safety Information

MSDS https://www.hit2lead.com/sds/4015723_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart