CH4102150745 ChemBridge Corporation

[4-chloro-2-(3,5-dimethyl-1H-pyrazol-1-yl)phenyl]boronic acid

  • MDL number MFCD08059990
  • Cas Number : 1287753-38-1
  • Molecular Weight : 250.4892

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4015832

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4015832_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart