CH4102150784 ChemBridge Corporation

(1-ethyl-1H-imidazol-5-yl)methanol

  • MDL number MFCD04114647
  • Cas Number : 215872-62-1
  • Molecular Weight : 126.1564

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4016572

Safety Information

MSDS https://www.hit2lead.com/sds/4016572_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart