CH4102150803 ChemBridge Corporation

(3-isobutylpiperidin-3-yl)methanol

  • MDL number MFCD08691611
  • Cas Number : 915922-54-2
  • Molecular Weight : 171.2798

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017651

Safety Information

MSDS https://www.hit2lead.com/sds/4017651_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart