CH4102150860 ChemBridge Corporation

1-cyclopentyl-1H-pyrazol-5-amine

  • MDL number MFCD03596775
  • Molecular Weight : 151.2089

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013058

Safety Information

MSDS https://www.hit2lead.com/sds/4013058_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart