CH4102150879 ChemBridge Corporation

5-(2-chlorophenyl)-2-furoic acid

  • MDL number MFCD02625435
  • Cas Number : 41019-43-6
  • Molecular Weight : 222.6245

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013146

Safety Information

MSDS https://www.hit2lead.com/sds/4013146_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart