CH4102150891 ChemBridge Corporation

2-(dimethylamino)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD03844445
  • Cas Number : 1005-28-3
  • Molecular Weight : 156.2055

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013292

Safety Information

MSDS https://www.hit2lead.com/sds/4013292_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart