CH4102150892 ChemBridge Corporation

2-pyrrolidin-1-yl-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD07021330
  • Cas Number : 900015-48-7
  • Molecular Weight : 182.2428

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013293

Safety Information

MSDS https://www.hit2lead.com/sds/4013293_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart