CH4102150900 ChemBridge Corporation

2-[4-(trifluoromethyl)phenyl]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD05865138
  • Cas Number : 144061-16-5
  • Molecular Weight : 273.231

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013405

Safety Information

MSDS https://www.hit2lead.com/sds/4013405_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart