CH4102150917 ChemBridge Corporation

5-(ethoxycarbonyl)-2,4-dimethyl-1H-pyrrole-3-carboxylic acid

  • MDL number MFCD00097477
  • Molecular Weight : 211.2145

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013658

Safety Information

MSDS https://www.hit2lead.com/sds/4013658_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart