CH4102150929 ChemBridge Corporation

3-[(5-methyl-1,3,4-thiadiazol-2-yl)thio]propanoic acid

  • MDL number MFCD04971051
  • Cas Number : 869943-40-8
  • Molecular Weight : 204.2699

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4013731

Safety Information

MSDS https://www.hit2lead.com/sds/4013731_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart