CH4102150968 ChemBridge Corporation

2,5-dihydrothiophene-3-sulfonyl chloride 1,1-dioxide

  • MDL number MFCD00196002
  • Cas Number : 112161-61-2
  • Molecular Weight : 216.6631

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011804

Safety Information

signalword Danger
hcodes 8
MSDS https://www.hit2lead.com/sds/4011804_sds.pdf
hstatements H314, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart