CH4102150998 ChemBridge Corporation

1-[2-(1H-imidazol-1-yl)phenyl]methanamine

  • MDL number MFCD06654837
  • Cas Number : 25373-55-1
  • Molecular Weight : 173.2144

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014237

Safety Information

MSDS https://www.hit2lead.com/sds/4014237_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart