CH4102151018 ChemBridge Corporation

3-methyl-1-propyl-1H-pyrazol-5-amine

  • MDL number MFCD05237221
  • Cas Number : 3524-34-3
  • Molecular Weight : 139.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4014323

Safety Information

MSDS https://www.hit2lead.com/sds/4014323_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart