CH4102151034 ChemBridge Corporation

1-(4-fluorobenzyl)-4-methyl-1H-pyrazol-5-amine

  • MDL number MFCD08691581
  • Cas Number : 1015845-96-1
  • Molecular Weight : 205.2315

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014364

Safety Information

MSDS https://www.hit2lead.com/sds/4014364_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart