CH4102151093 ChemBridge Corporation

(4-pyridin-2-ylbutyl)amine

  • MDL number MFCD07785855
  • Cas Number : 34974-00-0
  • Molecular Weight : 150.2209

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4014928

Safety Information

MSDS https://www.hit2lead.com/sds/4014928_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart