CH4102151096 ChemBridge Corporation

1-(1-ethylpiperidin-2-yl)methanamine

  • MDL number MFCD08059956
  • Cas Number : 37782-46-0
  • Molecular Weight : 142.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1-ethyl-2-piperidinyl)methylamine
Inchi Code 1S/C8H18N2/c1-2-10-6-4-3-5-8(10)7-9/h8H,2-7,9H2,1H3
Inchi Key LEKNBLYGRWKNSE-UHFFFAOYSA-N

Description

Purity 95%
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4014964

Safety Information

MSDS https://www.hit2lead.com/sds/4014964_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart