CH4102151136 ChemBridge Corporation

(tetrahydrofuran-2-ylmethoxy)acetic acid

  • MDL number MFCD07643288
  • Cas Number : 854702-18-4
  • Molecular Weight : 160.1678

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011117

Safety Information

MSDS https://www.hit2lead.com/sds/4011117_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart