CH4102151153 ChemBridge Corporation

2-(pyrrolidin-1-ylmethyl)piperidine

  • MDL number MFCD03426449
  • Cas Number : 100158-63-2
  • Molecular Weight : 168.2792

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011154

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4011154
Availability details for
Quantity
  • Availability
  • Add to Cart