CH4102151157 ChemBridge Corporation

4-(piperidin-2-ylmethyl)morpholine

  • MDL number MFCD05215079
  • Cas Number : 81310-58-9
  • Molecular Weight : 184.2786

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form liquid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4011155

Safety Information

MSDS https://www.hit2lead.com/sds/4011155_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart