CH4102151191 ChemBridge Corporation

5-(1H-pyrazol-5-yl)-2-furaldehyde

  • MDL number MFCD08059993
  • Cas Number : 1015939-90-8
  • Molecular Weight : 162.1454

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4015886

Safety Information

MSDS https://www.hit2lead.com/sds/4015886_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart