CH4102151210 ChemBridge Corporation

2-(1H-tetrazol-5-yl)benzoic acid

  • MDL number MFCD03308576
  • Cas Number : 13947-58-5
  • Molecular Weight : 190.1588

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012333

Safety Information

MSDS https://www.hit2lead.com/sds/4012333_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart