CH4102151238 ChemBridge Corporation

5-phenyl-4-propyl-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD05815986
  • Cas Number : 26029-08-3
  • Molecular Weight : 219.306

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012396

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4012396_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart