CH4102151240 ChemBridge Corporation

5-(3-methylphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD00618579
  • Cas Number : 75218-27-8
  • Molecular Weight : 191.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin RU
Product Link https://www.hit2lead.com/building-blocks/4012398

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4012398_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart