CH4102151244 ChemBridge Corporation

3-(3-formyl-1H-indol-1-yl)propanamide

  • MDL number MFCD04219232
  • Cas Number : 61922-01-8
  • Molecular Weight : 216.2359

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4016425

Safety Information

MSDS https://www.hit2lead.com/sds/4016425_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart