CH4102151248 ChemBridge Corporation

5-(3-methoxyphenyl)-4H-1,2,4-triazole-3-thiol

  • MDL number MFCD08691554
  • Cas Number : 419540-45-7
  • Molecular Weight : 207.2523

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012399

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4012399_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart