CH4102151257 ChemBridge Corporation

5-(2-methoxyphenyl)-1,3,4-oxadiazole-2-thiol

  • MDL number MFCD04614701
  • Cas Number : 69844-25-3
  • Molecular Weight : 208.237

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012409

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/sds/4012409_sds.pdf
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart