CH4102151280 ChemBridge Corporation

(3-chloro-2-morpholin-4-ylphenyl)amine

  • MDL number MFCD03821863
  • Cas Number : 58785-07-2
  • Molecular Weight : 212.676

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012422

Safety Information

MSDS https://www.hit2lead.com/sds/4012422_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart