CH4102151292 ChemBridge Corporation

3-chloro-1H-1,2,4-triazole

  • MDL number MFCD00128812
  • Cas Number : 6818-99-1
  • Molecular Weight : 103.5104

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017309

Safety Information

signalword Warning
MSDS https://www.hit2lead.com/building-blocks/4017309
hstatements H302, H315, H319, H335.
Availability details for
Quantity
  • Availability
  • Add to Cart