CH4102151307 ChemBridge Corporation

(4-isobutoxyphenyl)amine

  • MDL number MFCD01009861
  • Molecular Weight : 165.2322

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4017585

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4017585
Availability details for
Quantity
  • Availability
  • Add to Cart