CH4102151311 ChemBridge Corporation

4-(pyridin-2-ylmethoxy)aniline

  • MDL number MFCD04971040
  • Cas Number : 102137-46-2
  • Molecular Weight : 200.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4012449

Safety Information

MSDS https://www.hit2lead.com/sds/4012449_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart